Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Immobilization of biomolecules to plasma polymerized pentafluorophenyl methacrylate.
Immobilization of biomolecules to plasma polymerized pentafluorophenyl methacrylate.
Biomacromolecules (2010-09-14)
Luis Duque, Bernhard Menges, Salvador Borros, Renate Förch
ABSTRACT
Thin films of plasma polymerized pentafluorophenyl methacrylate (pp-PFM) offer highly reactive ester groups throughout the structure of the film that allow for subsequent reactions with different aminated reagents and biological molecules. The present paper follows on from previous work on the plasma deposition of pentafluorophenyl methacrylate (PFM) for optimum functional group retention (Francesch, L.; Borros, S.; Knoll, W.; Foerch, R. Langmuir 2007, 23, 3927) and reactivity in aqueous solution (Duque, L.; Queralto, N.; Francesch, L.; Bumbu, G. G.; Borros, S.; Berger, R.; Förch, R. Plasma Process. Polym. 2010, accepted for publication) to investigate the binding of a biologically active peptide known to induce cellular adhesion (IKVAV) and of biochemically active proteins such as BSA and fibrinogen. Analyses of the films and of the immobilization of the biomolecules were carried out using infrared reflection absorption spectroscopy (IRRAS), X-ray photoelectron spectroscopy (XPS), and atomic force microscopy (AFM). The attachment of the biomolecules on pulsed plasma polymerized pentafluorophenyl methacrylate was monitored using surface plasmon resonance spectroscopy (SPR). SPR analysis confirmed the presence of immobilized biomolecules on the plasma polymer and was used to determine the mass coverage of the peptide and proteins adsorbed onto the films. The combined analysis of the surfaces suggests the covalent binding of the peptide and proteins to the surface of the pp-PFM.
Sigma-Aldrich
Pentafluorophenyl methacrylate, contains MEHQ as inhibitor, 95%