Merck
A charge-switched nano-sized polymeric carrier for protein delivery.
International journal of pharmaceutics (2010-03-20)
Bo Reum Lee, Kyung Taek Oh, Hye Jung Baik, Yu Seok Youn, Eun Seong Lee
ABSTRACT
A novel synthetic nanocomplex was constructed from glycol chitosan (GCS) grafted with 2,3-dimethylmaleic anhydride (DMA) (denoted as 'GCD' hereafter) and lysozyme (isoelectric point=10.9) as a model protein. This is a core-shell supramolecular assemble formed through electrostatic interactions between anionic GCD and cationic lysozyme at a pH 7.4. The pH-sensitivity of the nanocomplexes originates from the dissociation of DMA block from GCD at a slightly acidic pH (i.e., pH 6.8), resulting in an increased electrostatic repulsion between cationic GCS and cationic lysozyme. This pH-induced charge switching of GCD provides a mechanism for triggered protein drug release from the nanocomplexes triggered by the small change in pH (pH 7.4-6.8).
Sigma-Aldrich
2,3-Dimethylmaleic anhydride, 98%