Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Use of the SPhos ligand to suppress racemization in arylpinacolboronate ester Suzuki couplings involving alpha-amino acids. Synthesis of biaryl derivatives of 4-hydroxyphenylglycine, tyrosine, and tryptophan.
Use of the SPhos ligand to suppress racemization in arylpinacolboronate ester Suzuki couplings involving alpha-amino acids. Synthesis of biaryl derivatives of 4-hydroxyphenylglycine, tyrosine, and tryptophan.
The Journal of organic chemistry (2009-10-30)
Mònica Prieto, Silvia Mayor, Paul Lloyd-Williams, Ernest Giralt
ABSTRACT
Alpha-amino acid derivatives, particularly those of phenylglycine, can suffer significant racemization in Suzuki couplings. When arylpinacolboronate esters are used as coupling partners this unwanted side reaction can be suppressed by the use of Pd(OAc)(2) as Pd(0) source, in the presence of Buchwald's SPhos ligand. The syntheses of biaryl amino acids of tyrosine, phenylglycine, and tryptophan, including Phg-Trp units similar to those found in the chloropeptin family of natural products, are reported.
Sigma-Aldrich
4-Hydroxy-L-phenylglycine, ≥99.0% (NT)