Merck
  • N-Propynyl analogs of beta-phenylethylidenehydrazines: synthesis and evaluation of effects on glycine, GABA, and monoamine oxidase.
N-Propynyl analogs of beta-phenylethylidenehydrazines: synthesis and evaluation of effects on glycine, GABA, and monoamine oxidase.
Bioorganic & medicinal chemistry (2008-08-12)
Erin M MacKenzie, Afshin Fassihi, Asghar Davood, Qiao-Hong Chen, Gillian Rauw, Gail Rauw, Edward E Knaus, Glen B Baker
ABSTRACT
A group of beta-phenylethylidenehydrazines possessing a variety of substituents (Me, OMe, Cl, F, and CF(3)) at the ortho-, meta-, or para-positions of the phenyl ring, in conjunction with either a N-bis-(2-propynyl) or a N-mono-(2-propynyl) moiety, were synthesized and compared to the novel neuroprotective drug beta-phenylethylidenehydrazine (PEH) with regard to their ability to inhibit the enzymes GABA-transaminase (GABA-T) and monoamine oxidase (MAO)-A and -B in vitro in brain tissue. Two of the analogs synthesized (mono- and bis-N-propynylPEH) were also studied exvivo in rats to compare their effects to those of PEH with regard to ability to inhibit GABA-T and MAO and to change brain levels of several important amino acids. Unlike PEH, none of the new drugs inhibited GABA-T in vitro at 10 or 100 microM, and all of the drugs (including PEH) were poor inhibitors (at 10 microM) of MAO-A and -B invitro. The two analogs studied exvivo inhibited GABA-T to a lesser extent than PEH, unlike PEH that did not elevate brain levels of GABA, and inhibited MAO-A and -B more potently than PEH. Interestingly, unlike PEH, the two analogs caused marked increases in brain levels of glycine; because of the current interest in drugs that increase glycine availability in the brain as potential antipsychotic drugs, these two analogs now warrant further investigation.
Sigma-Aldrich
Propyne, ≥99%
Sigma-Aldrich
Propyne, ≥97%