Merck
  • Genotoxic effects of PCB 52 and PCB 77 on cultured human peripheral lymphocytes.
Genotoxic effects of PCB 52 and PCB 77 on cultured human peripheral lymphocytes.
Mutation research (2008-06-25)
Suleyman Sandal, Bayram Yilmaz, David O Carpenter
ABSTRACT
Polychlorinated biphenyls (PCBs) are known to be carcinogenic, but the mechanisms of this action are uncertain. Most, but not all, studies have concluded that PCBs are not directly mutagenic, and that much if not all of the carcinogenic activity resides in the fraction of the PCB mixture that contains congeners with dioxin-like activity. The present study was designed to determine genotoxic effects of an ortho-substituted, non-coplanar congener, 2,2',5,5'-tetrachlorobiphenyl (PCB 52), and a non-ortho-substituted coplanar congener with dioxin-like activity, 3,3',4,4'-tetrachlorobiphenyl (PCB 77) on cultured human peripheral lymphocytes. DNA damage was assessed by use of the comet assay (alkaline single-cell gel electrophoresis). After cell cultures were prepared, test groups were treated with different concentrations of PCB 52 (0.2 and 1 microM) and PCB 77 (1 and 10 microM) for 1h at 37 degrees C in a humidified carbon dioxide incubator, and compared to a DMSO vehicle control group. The cells were visually classified into four categories on the basis of extent of migration such as undamaged (UD), low damage (LD), moderate damage (MD) and high damage (HD). The highest concentration of PCBs 52 and 77 significantly increased DNA breakage in human lymphocytes (p<0.001). Our results indicate that both the non-coplanar PCB 52 and coplanar PCB 77 cause DNA damage, and that the ortho-substituted congener was significantly more potent than the dioxin-like coplanar congener.
Product Description
2,2′,5,5′-Tetrachlorobiphenyl (IUPAC No. 52), BCR®, certified reference material
Supelco
PCB No 52, analytical standard