Merck
  • Laboratory study highlights the key influences of stormwater sediment thickness and bioturbation by tubificid worms on dynamics of nutrients and pollutants in stormwater retention systems.
Laboratory study highlights the key influences of stormwater sediment thickness and bioturbation by tubificid worms on dynamics of nutrients and pollutants in stormwater retention systems.
Chemosphere (2008-03-11)
F Mermillod-Blondin, G Nogaro, F Vallier, J Gibert
ABSTRACT
In urban area, the accumulation of polluted stormwater sediments (SWS) in retention ponds may be a source of dissolved pollutants and nutrients for the aquatic ecosystems. Our objective was to quantify the influence of the thickness of SWS layer and the occurrence of tubificid worms on organic matter processing (O(2) uptake and fluxes of NH(4)(+), NO(3)(-), PO(4)(3-), and dissolved organic carbon between sediment and water), releases of 17 PAHs and 4 heavy metals, and microbial characteristics. Results showed that oxidation of SWS organic matter (O(2) and NO(3)(-) uptakes) and releases of nutrients were significantly increased by the quantity of accumulated SWS and the worm bioturbation. Releases of acenaphtene and naphthalene from sediments were significantly increased by the thickness of the SWS layer. In contrast, tubificid worms did not promote the mobilization of pollutants. In conclusion, biological activities and stormwater sediment characteristics need to be assessed to quantify the fate of pollutants and nutrients in stormwater retention ponds.
Sigma-Aldrich
Acenaphthene, 99%
Supelco
Acenaphthene, analytical standard
Supelco
Acenaphthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland