Merck
  • Processes affecting the dissipation of the herbicide isoxaflutole and its diketonitrile metabolite in agricultural soils under field conditions.
Processes affecting the dissipation of the herbicide isoxaflutole and its diketonitrile metabolite in agricultural soils under field conditions.
Journal of agricultural and food chemistry (2007-09-21)
Sharon K Papiernik, Scott R Yates, William C Koskinen, Brian Barber
ABSTRACT
Two-year field dissipation studies were conducted in three soil types in Minnesota to examine the processes affecting the dissipation of the herbicide isoxaflutole and its phytotoxic diketonitrile metabolite (DKN) under relatively cool, wet soil conditions. Plots of cuphea were treated with isoxaflutole and potassium bromide, a nonsorbed, nondegraded tracer. Replicate soil cores were collected six times during the growing season to a depth of 1 m, and the bromide or herbicide concentration was measured in each of five depth increments. The dissipation half-life (DT50) of isoxaflutole + DKN was 8-18 days in each soil. Bromide and herbicide concentrations were low at depths >40 cm throughout the study, and herbicide concentrations in soil 100 days after application were usually undetectable. Simulation modeling using Hydrus-1D for the loam soil suggested that plant uptake was an important mechanism of dissipation.
Product Description
Supelco
Isoxaflutole, PESTANAL®, analytical standard