Merck
  • Crystal structures of Fe2+ dioxygenase superoxo, alkylperoxo, and bound product intermediates.
Crystal structures of Fe2+ dioxygenase superoxo, alkylperoxo, and bound product intermediates.
Science (New York, N.Y.) (2007-04-21)
Elena G Kovaleva, John D Lipscomb
ABSTRACT
We report the structures of three intermediates in the O2 activation and insertion reactions of an extradiol ring-cleaving dioxygenase. A crystal of Fe2+-containing homoprotocatechuate 2,3-dioxygenase was soaked in the slow substrate 4-nitrocatechol in a low O2 atmosphere. The x-ray crystal structure shows that three different intermediates reside in different subunits of a single homotetrameric enzyme molecule. One of these is the key substrate-alkylperoxo-Fe2+ intermediate, which has been predicted, but not structurally characterized, in an oxygenase. The intermediates define the major chemical steps of the dioxygenase mechanism and point to a general mechanistic strategy for the diverse 2-His-1-carboxylate enzyme family.
Sigma-Aldrich
4-Nitrocatechol, ≥96.0%