Merck
  • An efficient carbonyl-alkene metathesis of bicyclic oxetanes: photoinduced electron transfer reduction of the Paternò-Büchi adducts from 2,3-dihydrofuran and aromatic aldehydes.
An efficient carbonyl-alkene metathesis of bicyclic oxetanes: photoinduced electron transfer reduction of the Paternò-Büchi adducts from 2,3-dihydrofuran and aromatic aldehydes.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2006-01-06)
Raúl Pérez-Ruiz, Miguel A Miranda, Ronald Alle, Klaus Meerholz, Axel G Griesbeck
ABSTRACT
The bicyclic oxetanes and resulting from photocycloaddition of aromatic aldehydes to 2,3-dihydrofuran, were efficiently cleaved by means of electron-transfer reduction, photoinduced by the electronically excited reductants 1-methoxynaphthalene (MN) and 2,7-dimethoxynaphthalene (DMN) in acetonitrile. The fluorescence quenching rates of DMN/MN by and were determined by static methods, the triplet quenching rates were determined by means of laser flash photolysis (LFP). The product analysis established a "photo-photo metathesis" where both cycloaddition and cycloreversion processes are induced by photochemical processes.
Sigma-Aldrich
2,3-Dihydrofuran, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mL
Please contact Customer Service for Availability
€35.00
250 mL
Please contact Customer Service for Availability
€43.10
1 L
Please contact Customer Service for Availability
€62.10