Merck
  • Synthesis of 1,2-dioxolanes by annulation reactions of peroxycarbenium ions with alkenes.
Synthesis of 1,2-dioxolanes by annulation reactions of peroxycarbenium ions with alkenes.
Organic letters (2005-10-08)
Armando Ramirez, K A Woerpel
ABSTRACT
[reaction: see text] The annulation reactions of alkenes with peroxycarbenium ions enable the synthesis of a variety of functionalizable 1,2-dioxolanes. Triethysilyl-protected peroxycarbenium ions proved to be optimal for the annulation reaction. Using this method, plakinic acid analogues can be synthesized in three steps from the corresponding ketone and alkene.
Sigma-Aldrich
Allyltrimethylsilane, 98%