- Highly efficient chemical kinetic resolution of bishomoallylic alcohols: synthesis of (R)-sulcatol.
Highly efficient chemical kinetic resolution of bishomoallylic alcohols: synthesis of (R)-sulcatol.
Organic letters (2004-09-10)
Shui-Ling Chen, Qi-Ying Hu, Teck-Peng Loh
PMID15355053
ABSTRACT
[reaction: see text] A highly efficient chemical kinetic resolution of bishomoallylic alcohols was developed when the alcohols underwent In(OTf)(3)-catalyzed 3,5-oxonium-ene-type cyclization with steroidal aldehyde 2. Consistently high enantiomeric excess (up to >99%) was obtained.