Merck
  • Hypoglycemic effects of a beta-agonist, Ro 16-8714, in streptozotocin-diabetic rats: decreased hepatic glucose production and increased glucose utilization in oxidative muscles.
Hypoglycemic effects of a beta-agonist, Ro 16-8714, in streptozotocin-diabetic rats: decreased hepatic glucose production and increased glucose utilization in oxidative muscles.
Metabolism: clinical and experimental (1992-02-01)
P Ferré, L Pénicaud, Y Hitier, M Meier, J Girard
ABSTRACT
Streptozotocin (STZ)-induced diabetic rats are glycosuric, hyperglycemic, hyperketonemic, overproduce glucose, and have a decreased glucose utilization in oxidative muscles. Treatment with a beta-agonist, Ro 16-8714, decreases the glycosuria, hyperglycemia, hyperketonemia, and hepatic glucose production. Tissue glucose utilization was unchanged, except in oxidative muscles, where it was increased.
Sigma-Aldrich
2-Amino-1-phenylethanol, 98%