Merck
  • Interactions of oxyphenbutazone with different cyclodextrins in aqueous medium and in the solid state.
Interactions of oxyphenbutazone with different cyclodextrins in aqueous medium and in the solid state.
Journal of pharmaceutical and biomedical analysis (2002-06-01)
M D Veiga, M Merino
ABSTRACT
The interactions between a nonsteroidal anti-inflammatory drugs, oxyphenbutazone (OPB), with two cyclodextrins, beta-cyclodextrin (beta-CD) and gamma-cyclodextrin (gamma-CD), have been studied in an aqueous medium and in the solid state. Differential scanning calorimetry, hot stage microscopy, thermogravimetric analysis and X-ray diffraction (XRD) powder have been the techniques used to characterize the interactions in the solid state. Although OPB forms inclusion compounds with beta- and gamma-CD in the aqueous medium, only the OPB/gamma-CD inclusion compound was obtained in the solid state by the kneading method. The XRD powder used at different temperatures has proven be a useful tool in characterizing the behaviour of these binary systems.
Sigma-Aldrich
Oxyphenbutazone, ≥98% (HPLC)