Merck
  • Study of adjuvant effect of model surfactants from the groups of alkyl sulfates, alkylbenzene sulfonates, alcohol ethoxylates and soaps.
Study of adjuvant effect of model surfactants from the groups of alkyl sulfates, alkylbenzene sulfonates, alcohol ethoxylates and soaps.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2000-10-19)
S K Clausen, S Sobhani, O M Poulsen, L K Poulsen, G D Nielsen
ABSTRACT
The sodium salts of representatives of anionic surfactants, dodecylbenzene sulfonate (SDBS), dodecyl sulfate (SDS) and coconut oil fatty acids, and a nonionic surfactant, dodecyl alcohol ethoxylate, were studied for adjuvant effect on the production of specific IgE antibodies in mice. The surfactants were injected subcutaneously (sc) in concentrations of 1000, 100, 10 or 1 mg/l, respectively, together with 1 microg of ovalbumin (OVA). In addition, groups of mice received OVA in saline (control group) or in Al(OH)(3) (positive adjuvant control group). After the primary immunization the mice were boosted up to three times with OVA (0.1 microg sc) in saline. OVA-specific IgE antibodies were determined by the heterologous mouse rat passive cutaneous anaphylaxis test. The results were confirmed by a specific ELISA method. After the first booster, the Al(OH)(3) group and the 10 mg/l SDS group showed a statistically significant increase in OVA specific IgE levels. After two boosters, a statistically significant suppression in OVA-specific IgE production occurred with SDS (1000 mg/l), SDBS (1000 and 100 mg/l), coconut soap (1000 mg/l) and the alcohol ethoxylate (10 mg/l). This study suggests that a limited number of surfactants possess an adjuvant effect whereas all surfactants at certain levels can suppress specific IgE production.
Product Description
Supelco
1-Phenyldodecane, analytical standard
Sigma-Aldrich
1-Phenyldodecane, 97%
Sigma-Aldrich
Genapol® X-100
Sigma-Aldrich
Genapol® X-080