Merck
  • Cyclopentenones, scaffolds for organic syntheses produced by the endophytic fungus mitosporic dothideomycete sp. LRUB20.
Cyclopentenones, scaffolds for organic syntheses produced by the endophytic fungus mitosporic dothideomycete sp. LRUB20.
Journal of natural products (2006-09-23)
Porntep Chomcheon, Nongluksna Sriubolmas, Suthep Wiyakrutta, Nattaya Ngamrojanavanich, Narongsak Chaichit, Chulabhorn Mahidol, Somsak Ruchirawat, Prasat Kittakoop
ABSTRACT
Two new natural products, 2-hydroxymethyl-3-methylcyclopent-2-enone (1) (synthetically known) and cis-2-hydroxymethyl-3-methylcyclopentanone (2), and a known compound, asterric acid (3), were isolated from the endophytic fungus mitosporic Dothideomycete sp. LRUB20, which was isolated from the stem of the Thai medicinal plant Leea rubra. Compound 2 was separated and identified in the form of its 2,4-dinitrophenylhydrazone derivative (5). Compounds 1, 3, and hydrazone 5 exhibited mild antimycobacterial activity, both with MIC values of 200 microg/mL. Compounds 1, 3, and 4 were inactive (at 50 microg/mL) toward Vero, KB, NCI-H187, and BC cell lines. Hydrazone 5 showed only mild cytotoxicity against the Vero cell line with an IC(50) value of 21.7 microg/mL; however, it was inactive toward KB, NCI-H187, and BC cell lines. Endophytic fungi may be a source for the production of building blocks for organic syntheses.
Sigma-Aldrich
Asterric acid, ≥95% (LC/MS-ELSD)