Merck
  • Controlling integrin specificity and stem cell differentiation in 2D and 3D environments through regulation of fibronectin domain stability.
Controlling integrin specificity and stem cell differentiation in 2D and 3D environments through regulation of fibronectin domain stability.
Biomaterials (2008-11-26)
Mikaël M Martino, Mayumi Mochizuki, Dominique A Rothenfluh, Sandra A Rempel, Jeffrey A Hubbell, Thomas H Barker
ABSTRACT
The extracellular matrix (ECM) exerts powerful control over many cellular phenomena, including stem cell differentiation. As such, design and modulation of ECM analogs to ligate specific integrin is a promising approach to control cellular processes in vitro and in vivo for regenerative medicine strategies. Although fibronectin (FN), a crucial ECM protein in tissue development and repair, and its RGD peptide are widely used for cell adhesion, the promiscuity with which they engage integrins leads to difficulty in control of receptor-specific interactions. Recent simulations of force-mediated unfolding of FN domains and sequences analysis of human versus mouse FN suggest that the structural stability of the FN's central cell-binding domains (FN III9-10) affects its integrin specificity. Through production of FN III9-10 variants with variable stabilities, we obtained ligands that present different specificities for the integrin alpha(5)beta(1) and that can be covalently linked into fibrin matrices. Here, we demonstrate the capacity of alpha(5)beta(1) integrin-specific engagement to influence human mesenchymal stem cell (MSC) behavior in 2D and 3D environments. Our data indicate that alpha(5)beta(1) has an important role in the control of MSC osteogenic differentiation. FN fragments with increased specificity for alpha(5)beta(1) versus alpha(v)beta(3) results in significantly enhanced osteogenic differentiation of MSCs in 2D and in a clinically relevant 3D fibrin matrix system, although attachment/spreading and proliferation were comparable with that on full-length FN. This work shows how integrin-dependant cellular interactions with the ECM can be engineered to control stem cell fate, within a system appropriate for both 3D cell culture and tissue engineering.
Sigma-Aldrich
Thrombin from bovine plasma, ≥60 NIH units/mg protein (biuret)
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, 600-2,000 NIH units/mg protein (biuret)
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, 40-500 NIH units/mg protein (biuret)
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%