Merck
  • The key role of IL-6-arginase cascade for inducing dendritic cell-dependent CD4(+) T cell dysfunction in tumor-bearing mice.
The key role of IL-6-arginase cascade for inducing dendritic cell-dependent CD4(+) T cell dysfunction in tumor-bearing mice.
Journal of immunology (Baltimore, Md. : 1950) (2012-12-19)
Yoshinori Narita, Hidemitsu Kitamura, Daiko Wakita, Kentaro Sumida, Kazutaka Masuko, Satoshi Terada, Kiichiroh Nakano, Takashi Nishimura
ABSTRACT
Evaluation of immune dysfunction during the tumor-bearing state is a critical issue in combating cancer. In this study, we initially found that IL-6, one of the cachectic factors, suppressed CD4(+) T cell-mediated immunity through downregulation of MHC class II by enhanced arginase activity of dendritic cells (DC) in tumor-bearing mice. We demonstrated that administration of Ab against IL-6R (anti-IL-6R mAb) greatly enhanced T cell responses and inhibited the growth of tumor in vivo. We also found that IL-6 upregulated the expression of arginase-1 and arginase activity of DC in vitro. Tumor-infiltrating CD11c(+) DC exhibited upregulated mRNA expression of arginase-1 but reduced expression of MHC class II in parallel with the increase in serum IL-6 levels at the late stage in tumor-bearing hosts. However, the administration of anti-IL-6R mAb into tumor-bearing mice inhibited both the downmodulation of MHC class II and the upregulation of arginase-1 mRNA levels in DC. Furthermore, we noted that N(ω)-hydroxy-L-arginine or L-arginine, an arginase-1 inhibitor, blocked the reduction in MHC class II levels on CD11c(+) DC during the tumor-bearing state. Finally, we demonstrated that the administration of N(ω)-hydroxy-L-arginine at the peritumor site significantly enhanced CD4(+) T cell responses and inhibited tumor growth. Thus, IL-6-mediated arginase activation and the subsequent reduction in MHC class II expression on DC appeared to be critical mechanisms for inducing dysfunction of the immune system in the tumor-bearing state. Blockade of the IL-6-arginase cascade is a promising tool to overcome the dysfunction of antitumor immunity in tumor-bearing hosts.
Sigma-Aldrich
L-Arginase from bovine liver, Protein ≥70 % by biuret, powder