- Nanomechanical properties of electrospun composite scaffolds based on polycaprolactone and hydroxyapatite.
Nanomechanical properties of electrospun composite scaffolds based on polycaprolactone and hydroxyapatite.
Fibrous nanocomposite scaffolds were electrospun from dispersions of hydroxyapatite nanoparticles (nanoHA) in polycaprolactone (PCL) with varying nanoHA contents (from 0% to 50% by weight). Such scaffolds were produced to mimic the nano-features of the extracellular matrix (ECM) for natural bone tissue regeneration. NanoHA was found to be well dispersed in the PCL fibers up to the addition of 30 wt%, whereas beads and agglomeration of HA particles was observed above this nanoHA concentration. The structural and morphological characterizations were evaluated by scanning electron microscopy (SEM), Fourier-Transform Infrared Spectroscopy (FTIR), and X-ray Diffraction (XRD). The average fiber diameter decreased with increased nanoHA concentration. The nanomechanical properties of the as-spun fibrous scaffolds as well as pressure-consolidated (pelletized) composites were evaluated by nanoindentation. Elastic modulus increased with increasing HA content, but was especially pronounced for 40-50% HA content where the indenter tip is more likely to probe agglomerated HA particles.