Merck
  • The typically disordered N-terminus of PKA can fold as a helix and project the myristoylation site into solution.
The typically disordered N-terminus of PKA can fold as a helix and project the myristoylation site into solution.
Biochemistry (2004-06-16)
Christine Breitenlechner, Richard A Engh, Robert Huber, Volker Kinzel, Dirk Bossemeyer, Michael Gassel
ABSTRACT
Protein kinases comprise the major enzyme family critically involved in signal transduction pathways; posttranslational modifications affect their regulation and determine signaling states. The prototype protein kinase A (PKA) possesses an N-terminal alpha-helix (Helix A) that is atypical for kinases and is thus a major distinguishing feature of PKA. Its physiological function may involve myristoylation at the N-terminus and modulation via phosphorylation at serine 10. Here we describe an unusual structure of an unmyristoylated PKA, unphosphorylated at serine 10, with a completely ordered N-terminus. Using standard conditions (e.g., PKI 5-24, ATP site ligand, MEGA-8), a novel 2-fold phosphorylated PKA variant showed the ordered N-terminus in a new crystal packing arrangement. Thus, the critical factor for structuring the N-terminus is apparently the absence of phosphorylation of Ser10. The flexibility of the N-terminus, its myristoylation, and the conformational dependence on the phosphorylation state are consistent with a functional role for myristoylation.
Sigma-Aldrich
N-Octanoyl-N-methylglucamine, ≥97% (GC)