Merck
  • Wnt5a-YAP signaling axis mediates mechanotransduction in cardiac myocytes and contributes to contractile dysfunction induced by pressure overload.
Wnt5a-YAP signaling axis mediates mechanotransduction in cardiac myocytes and contributes to contractile dysfunction induced by pressure overload.
iScience (2023-07-17)
Hiroshi Kishimoto, Masayoshi Iwasaki, Kensaku Wada, Keita Horitani, Osamu Tsukamoto, Kenta Kamikubo, Seitaro Nomura, Shinji Matsumoto, Takeshi Harada, Daisuke Motooka, Daisuke Okuzaki, Seiji Takashima, Issei Komuro, Akira Kikuchi, Ichiro Shiojima
ABSTRACT
Non-canonical Wnt signaling activated by Wnt5a/Wnt11 is required for the second heart field development in mice. However, the pathophysiological role of non-canonical Wnt signaling in the adult heart has not been fully elucidated. Here we show that cardiomyocyte-specific Wnt5a knockout mice exhibit improved systolic function and reduced expression of mechanosensitive genes including Nppb when subjected to pressure overload. In cultured cardiomyocytes, Wnt5a knockdown reduced Nppb upregulation induced by cyclic cell stretch. Upstream analysis revealed that TEAD1, a transcription factor that acts with Hippo pathway co-activator YAP, was downregulated both in vitro and in vivo by Wnt5a knockdown/knockout. YAP nuclear translocation was induced by cell stretch and attenuated by Wnt5a knockdown. Wnt5a knockdown-induced Nppb downregulation during cell stretch was rescued by Hippo inhibition, and the rescue effect was canceled by knockdown of YAP. These results collectively suggest that Wnt5a-YAP signaling axis mediates mechanotransduction in cardiomyocytes and contributes to heart failure progression.
Sigma-Aldrich
XMU-MP-1 hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Tamoxifen, ≥99%