Merck
  • Carbonate apatite increases gene expression of osterix and bone morphogenetic protein 2 in the alveolar ridge after socket grafting.
Carbonate apatite increases gene expression of osterix and bone morphogenetic protein 2 in the alveolar ridge after socket grafting.
Journal of oral science (2023-11-27)
Tadashi Kitazawa, Hideki Takai, Tetsuro Kono, Hiroyuki Okada, Yorimasa Ogata
ABSTRACT
After tooth extraction, preservation of the alveolar ridge by socket grafting attenuates bone resorption. Runt-related transcription factor 2 (RUNX2) and SP7/Osterix (OSX) are transcription factors playing an important role in osteoblast differentiation. The purpose of this study was to evaluate the effects of carbonate apatite (CO3Ap) on osteoblast-related gene and protein expression after socket grafting. Alveolar bone and new bone after CO3Ap grafting were collected at the time of implant placement. Levels of mRNA for RUNX2, SP7/OSX, bone morphogenetic protein 2 (BMP2), BMP7 and platelet derived growth factor B were determined by real-time PCR. Immunostaining was performed using antibodies against RUNX2, SP7/OSX, vimentin and cytokeratin. To evaluate bone resorption rates, cone-beam CT (CBCT) imaging was performed after socket grafting and before implant placement. CBCT imaging showed that the average degree of bone resorption at the CO3Ap graft site was 7.15 ± 3.79%. At the graft sites, levels of SP7/OSX and BMP2 mRNA were significantly increased. Replacement of CO3Ap with osteoid was evident histologically, and in the osteoid osteoblast-like cells were stained for SP7/OSX and vimentin. These results show that gene expression of both SP7/OSX and BMP2 can be induced by CO3Ap, suggesting that increased expression of SP7/OSX and vimentin may be involved in the BMP pathway.
Sigma-Aldrich
Anti-Cytokeratin Epithelial Antibody, clone AE1, clone AE1, Chemicon®, from mouse