Merck
  • Combined Inter- and Intramolecular Charge-Transfer Processes for Highly Efficient Fluorescent Organic Light-Emitting Diodes with Reduced Triplet Exciton Quenching.
Combined Inter- and Intramolecular Charge-Transfer Processes for Highly Efficient Fluorescent Organic Light-Emitting Diodes with Reduced Triplet Exciton Quenching.
Advanced materials (Deerfield Beach, Fla.) (2017-03-02)
Chang-Ki Moon, Katsuaki Suzuki, Katsuyuki Shizu, Chihaya Adachi, Hironori Kaji, Jang-Joo Kim
ABSTRACT
Inter- and intramolecular charge-transfer processes are combined using an exciplex-forming host and a thermally activated delayed fluorescent dopant, for fabricating efficient fluorescent organic light-emitting diodes along with the reduced efficiency roll-off at high current densities. Extra conversion on the host from triplet exciplexes to singlet exciplexes followed by energy transfer to the dopant reduces population of triplet excitons on dopant molecules, thereby reducing the triplet exciton annihilations at high current densities.
Sigma-Aldrich
TcTa, ≥99% (HPLC)