Merck
  • Single-cell RNA sequencing reveals the epithelial cell heterogeneity and invasive subpopulation in human bladder cancer.
Single-cell RNA sequencing reveals the epithelial cell heterogeneity and invasive subpopulation in human bladder cancer.
International journal of cancer (2021-09-05)
Huadong Lai, Xiaomu Cheng, Qiang Liu, Wenqin Luo, Mengyao Liu, Man Zhang, Juju Miao, Zhongzhong Ji, Guan Ning Lin, Weichen Song, Lianhua Zhang, Juanjie Bo, Guoliang Yang, Jia Wang, Wei-Qiang Gao
ABSTRACT
Bladder cancer represents a highly heterogeneous disease characterized by distinct histological, molecular and clinical phenotypes, and a detailed analysis of tumor cell invasion and crosstalks within bladder tumor cells has not been determined. Here, we applied droplet-based single-cell RNA sequencing (scRNA-seq) to acquire transcriptional profiles of 36 619 single cells isolated from seven patients. Single cell transcriptional profiles matched well with the pathological basal/luminal subtypes. Notably, in T1 tumors diagnosed as luminal subtype, basal cells displayed characteristics of epithelial-mesenchymal transition (EMT) and mainly located at the tumor-stromal interface as well as micrometastases in the lamina propria. In one T3 tumor, muscle-invasive tumor showed significantly higher expression of cancer stem cell markers SOX9 and SOX2 than the primary tumor. We additionally analyzed communications between tumor cells and demonstrated its relevance to basal/luminal phenotypes. Overall, our single-cell study provides a deeper insight into the tumor cell heterogeneity associated with bladder cancer progression.
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit