Merck
  • A Novel Muscle Atrophy Mechanism: Myocyte Degeneration Due to Intracellular Iron Deprivation.
A Novel Muscle Atrophy Mechanism: Myocyte Degeneration Due to Intracellular Iron Deprivation.
Cells (2022-09-24)
Dae Keun Suh, Won-Young Lee, Woo Jin Yeo, Bong Soo Kyung, Koo Whang Jung, Hye Kyung Seo, Yong-Soo Lee, Dong Won Suh
ABSTRACT
Muscle atrophy is defined as the progressive degeneration or shrinkage of myocytes and is triggered by factors such as aging, cancer, injury, inflammation, and immobilization. Considering the total amount of body iron stores and its crucial role in skeletal muscle, myocytes may have their own iron regulation mechanism. Although the detrimental effects of iron overload or iron deficiency on muscle function have been studied, the molecular mechanism of iron-dependent muscle atrophy has not been elucidated. Using human muscle tissues and in the mouse rotator cuff tear model, we confirmed an association between injury-induced iron depletion in myocytes and muscle atrophy. In differentiated C2C12 myotubes, the effects of iron deficiency on myocytes and the molecular mechanism of muscle atrophy by iron deficiency were evaluated. Our study revealed that the lower iron concentration in injured muscle was associated with the upregulation of ferroportin, an iron exporter that transports iron out of cells. Ferroportin expression was increased by hypoxia-inducible factor 1α (HIF1α), which is activated by muscle injury, and its expression is controlled by HIF1 inhibitor treatment. Iron deprivation caused myocyte loss and a marked depletion of mitochondrial membrane potential leading to muscle atrophy, together with increased levels of myostatin, the upstream regulator of atrogin1 and muscle RING-finger protein-1 (MuRF1). Myostatin expression under iron deficiency was mediated by an orphan nuclear receptor, dosage-sensitive sex reversal-adrenal hypoplasia congenita critical region on the X chromosome (DAX1).