Merck
  • Zinc Supplementation Enhances the Pro-Death Function of UPR in Lymphoma Cells Exposed to Radiation.
Zinc Supplementation Enhances the Pro-Death Function of UPR in Lymphoma Cells Exposed to Radiation.
Biology (2022-01-22)
Roberta Gonnella, Luisa Guttieri, Maria Saveria Gilardini Montani, Roberta Santarelli, Erica Bassetti, Gabriella D'Orazi, Mara Cirone
ABSTRACT
We have previously shown that Zinc supplementation triggered ER stress/UPR in cancer cells undergoing treatment by genotoxic agents, reactivated wtp53 in cancer cells harboring mutant p53 (mutp53) and potentiated the activity of wtp53 in those carrying wtp53. In this study, we used Zinc chloride alone or in combination with 2 Gy radiation to treat Primary Effusion Lymphoma (PEL) cells, an aggressive B-cell lymphoma associated with KSHV that harbors wt or partially functioning p53. We found that Zinc triggered a mild ER stress/UPR in these lymphoma cells and activated ERK1/2, molecule known to sustain cell survival in the course of UPR activation. In combination with radiations, Zinc triggered a stronger p53 activation that counteracted its mediated ERK1/2 phosphorylation, further upregulating the UPR molecule CHOP and promoting cell death. These data suggest that Zinc supplementation could be a promising strategy to reduce the doses of radiation and possibly of other DNA-damaging agents to obtain an efficient capacity to induce lymphoma cell death.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Pifithrin-α, A cell-permeable chemical inhibitor of p53.
Sigma-Aldrich
PD 98,059, solid