Merck
  • Angiotensin-(1-7) exerts a protective action in a rat model of ventilator-induced diaphragmatic dysfunction.
Angiotensin-(1-7) exerts a protective action in a rat model of ventilator-induced diaphragmatic dysfunction.
Intensive care medicine experimental (2019-01-20)
Vanessa Zambelli, Anna Sigurtà, Laura Rizzi, Letizia Zucca, Paolo Delvecchio, Elena Bresciani, Antonio Torsello, Giacomo Bellani
ABSTRACT
Ventilator-induced diaphragmatic dysfunction (VIDD) is a common event during mechanical ventilation (MV) leading to rapid muscular atrophy and contractile dysfunction. Recent data show that renin-angiotensin system is involved in diaphragmatic skeletal muscle atrophy after MV. In particular, angiotensin-II can induce marked diaphragm muscle wasting, whereas angiotensin-(1-7) (Ang-(1-7)) could counteract this activity. This study was designed to evaluate the effects of the treatment with Ang-(1-7) in a rat model of VIDD with neuromuscular blocking agent infusion. Moreover, we studied whether the administration of A-779, an antagonist of Ang-(1-7) receptor (Mas), alone or in combination with PD123319, an antagonist of AT2 receptor, could antagonize the effects of Ang-(1-7). Sprague-Dawley rats underwent prolonged MV (8 h), while receiving an iv infusion of sterile saline 0.9% (vehicle) or Ang-(1-7) or Ang-(1-7) + A-779 or Ang-(1-7) + A-779 + PD123319. Diaphragms were collected for ex vivo contractility measurement (with electric stimulation), histological analysis, quantitative real-time PCR, and Western blot analysis. MV resulted in a significant reduction of diaphragmatic contractility in all groups of treatment. Ang-(1-7)-treated rats showed higher muscular fibers cross-sectional area and lower atrogin-1 and myogenin mRNA levels, compared to vehicle treatment. Treatment with the antagonists of Mas and Ang-II receptor 2 (AT2R) caused a significant reduction of muscular contractility and an increase of atrogin-1 and MuRF-1 mRNA levels, not affecting the cross-sectional fiber area and myogenin mRNA levels. Systemic Ang-(1-7) administration during MV exerts a protective role on the muscular fibers of the diaphragm preserving muscular fibers anatomy, and reducing atrophy. The involvement of Mas and AT2R in the mechanism of action of Ang-(1-7) still remains controversial.
Sigma-Aldrich
Angiotensin Fragment 1-7 acetate salt hydrate, ≥90% (HPLC), powder