- Requirement for activin A and transforming growth factor--beta 1 pro-regions in homodimer assembly.
Requirement for activin A and transforming growth factor--beta 1 pro-regions in homodimer assembly.
Science (New York, N.Y.) (1990-03-16)
A M Gray, A J Mason
PMID2315700
ABSTRACT
Many proteins are initially synthesized as part of a large precursor. The role of the pro-region in the biosynthesis of transforming growth factor--beta 1 (TGF-beta 1) and activin A, two structurally related disulfide-linked homodimers synthesized as large precursors, was studied. Vectors that expressed either the pro-region or the mature regions of these molecules were used in complementation experiments, only when the pro-region was coexpressed with the mature region did intracellular dimerization and secretion of biologically active homodimers occur. The pro-regions of activin A and TGF-beta 1, therefore, aid the folding, disulfide bond formation, and export of their respective homodimers.
Sigma-Aldrich
Anti-Pro GDF11 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution