Merck
  • Development and Characterization of a Porcine Mitral Valve Scaffold for Tissue Engineering.
Development and Characterization of a Porcine Mitral Valve Scaffold for Tissue Engineering.
Journal of cardiovascular translational research (2017-05-04)
M Granados, L Morticelli, S Andriopoulou, P Kalozoumis, M Pflaum, P Iablonskii, B Glasmacher, M Harder, J Hegermann, C Wrede, I Tudorache, S Cebotari, A Hilfiker, A Haverich, Sotirios Korossis
ABSTRACT
Decellularized scaffolds represent a promising alternative for mitral valve (MV) replacement. This work developed and characterized a protocol for the decellularization of whole MVs. Porcine MVs were decellularized with 0.5% (w/v) SDS and 0.5% (w/v) SD and sterilized with 0.1% (v/v) PAA. Decellularized samples were seeded with human foreskin fibroblasts and human adipose-derived stem cells to investigate cellular repopulation and infiltration, and with human colony-forming endothelial cells to investigate collagen IV formation. Histology revealed an acellular scaffold with a generally conserved histoarchitecture, but collagen IV loss. Following decellularization, no significant changes were observed in the hydroxyproline content, but there was a significant reduction in the glycosaminoglycan content. SEM/TEM analysis confirmed cellular removal and loss of some extracellular matrix components. Collagen and elastin were generally preserved. The endothelial cells produced newly formed collagen IV on the non-cytotoxic scaffold. The protocol produced acellular scaffolds with generally preserved histoarchitecture, biochemistry, and biomechanics.
Sigma-Aldrich
RPMI-1640 Medium, With L-alanyl-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture