Merck
Fluorescence based strategies for genetic analysis.
Chemical communications (Cambridge, England) (2005-12-17)
Rohan T Ranasinghe, Tom Brown
ABSTRACT
Synthetic chemistry has been central to the design of modern methods of genetic analysis. In this article, we discuss the underlying chemistry and biophysical principles that have been used in the development of robust methods for the analysis of DNA in the diagnostic laboratory.