Merck
  • Role of 657del5 NBN mutation and 7p12.2 (IKZF1), 9p21 (CDKN2A), 10q21.2 (ARID5B) and 14q11.2 (CEBPE) variation and risk of childhood ALL in the Polish population.
Role of 657del5 NBN mutation and 7p12.2 (IKZF1), 9p21 (CDKN2A), 10q21.2 (ARID5B) and 14q11.2 (CEBPE) variation and risk of childhood ALL in the Polish population.
Leukemia research (2011-09-06)
Agata Pastorczak, Patryk Górniak, Amy Sherborne, Fay Hosking, Joanna Trelińska, Monika Lejman, Tomasz Szczepański, Maciej Borowiec, Wojciech Fendler, Jerzy Kowalczyk, Richard S Houlston, Wojciech Młynarski
ABSTRACT
Recent studies have shown that SNPs mapping to 7p12.2 (IKZF1), 9p21 (CDKN2A), 10q21.2 (ARID5B), and 14q11.2 (CEBPE) and carrier status for recessively inherited Nijmegen Breakage syndrome (NBS) influence childhood acute lymphoblastic leukemia (ALL) risk. To examine these relationship, we analysed 398 ALL cases and 731 controls from Poland. Statistically significant association between genotype at 7p12.2 (IKZF1), 10q21.2 (ARID5B) and the NBS associated locus, 8q21.3 (NBN) and ALL risk was found; odds ratios (ORs), 1.34 (P=0.002), 1.33 (P=0.003), and 1325.21 (P=0.0028), respectively. These data provide further insights into the biological basis of ALL highlighting the existence of both common and rare disease susceptibility variants.