Merck
  • Nanocluster-Based Ultralow-Temperature Driven Oxide Gate Dielectrics for High-Performance Organic Electronic Devices.
Nanocluster-Based Ultralow-Temperature Driven Oxide Gate Dielectrics for High-Performance Organic Electronic Devices.
Materials (Basel, Switzerland) (2020-12-11)
Jeong-Wan Jo, Jingu Kang, Kyung-Tae Kim, Seung-Han Kang, Jae-Cheol Shin, Seung Beom Shin, Yong-Hoon Kim, Sung Kyu Park
ABSTRACT
The development of novel dielectric materials with reliable dielectric properties and low-temperature processibility is crucial to manufacturing flexible and high-performance organic thin-film transistors (OTFTs) for next-generation roll-to-roll organic electronics. Here, we investigate the solution-based fabrication of high-k aluminum oxide (Al2O3) thin films for high-performance OTFTs. Nanocluster-based Al2O3 films fabricated by highly energetic photochemical activation, which allows low-temperature processing, are compared to the conventional nitrate-based Al2O3 films. A wide array of spectroscopic and surface analyses show that ultralow-temperature photochemical activation (<60 °C) induces the decomposition of chemical impurities and causes the densification of the metal-oxide film, resulting in a highly dense high-k Al2O3 dielectric layer from Al-13 nanocluster-based solutions. The fabricated nanocluster-based Al2O3 films exhibit a low leakage current density (<10-7 A/cm2) at 2 MV/cm and high dielectric breakdown strength (>6 MV/cm). Using this dielectric layer, precisely aligned microrod-shaped 2,7-dioctyl[1]benzothieno [3,2-b][1] benzothiophene (C8-BTBT) single-crystal OTFTs were fabricated via solvent vapor annealing and photochemical patterning of the sacrificial layer.
Sigma-Aldrich
C8-BTBT, ≥99% (HPLC)