Merck
  • A comparative study of long interspersed element-1 protein immunoreactivity in cutaneous malignancies.
A comparative study of long interspersed element-1 protein immunoreactivity in cutaneous malignancies.
BMC cancer (2020-06-20)
Mohammad Ali Zolfaghari, Abbas Karimi, Elham Kalantari, Alireza Korourian, Alireza Ghanadan, Kambiz Kamyab, Nafiseh Esmaili, Amir Nader Emami Razavi, Zahra Madjd
ABSTRACT
Skin cancer is the most common cancer worldwide and commonly classified into malignant melanoma (MM) and Nonmelanoma skin cancers (NMSCs), which mainly include basal cell carcinoma (BCC) and squamous cell carcinoma (SCC). The extent to which Long Interspersed Element-1 (LINE-1, L1) ORF1p is expressed in cutaneous malignancies remains to be evaluated. This study aimed to assess LINE-1 ORF1p immunoreactivity in various skin cancer subtypes. The expression level of LINE-1 ORF1p was evaluated in 95 skin cancer specimens comprising 36 (37.9%) BCC, 28 (29.5%) SCC, and 31 (32.6%) melanoma using the tissue microarray (TMA) technique. Then the association between expression of LINE-1 encoded protein and clinicopathological parameters was analyzed. We showed that LINE-1 ORF1p expression level was substantially higher in BCC and SCC patients compared with melanoma samples (p < 0.001). BCC cases had a higher LINE-1 histochemical score (H-score) compared with SCC cases (p = 0.004). In SCC samples, a lower level of LINE-1 ORF1p expression was associated with age younger than the mean (p = 0.041). At the same time, no significant correlation was found between LINE-1 ORF1p expression and other clinicopathological parameters (all p > 0.05). According to our observation, LINE-1 ORF1p immunoreactivity in various skin tumor subtypes extends previous studies of LINE-1 expression in different cancers. LINE-1ORF1p overexpression in NMSCs compared with MM can be considered with caution as a tumor-specific antigen for NMSCs.
Sigma-Aldrich
Anti-LINE-1 ORF1p Antibody, clone 4H1, clone 4H1, from mouse