Merck
  • Increases in c-Src expression level and activity do not promote the growth of human colorectal carcinoma cells in vitro and in vivo.
Increases in c-Src expression level and activity do not promote the growth of human colorectal carcinoma cells in vitro and in vivo.
Neoplasia (New York, N.Y.) (2006-11-30)
Arkadiusz Welman, Christopher Cawthorne, Lourdes Ponce-Perez, Jane Barraclough, Sarah Danson, Stephen Murray, Jeff Cummings, Terry D Allen, Caroline Dive
ABSTRACT
The levels and activity of c-Src in colorectal cancer cells increase steadily during the course of colorectal carcinogenesis and are most highly elevated in advanced metastatic disease. However, the effects of increases in c-Src activity on the proliferation of colorectal cancer cells during early and late stages of tumorigenesis remain elusive. To study the consequences of increases in c-Src levels and activity on the growth of colorectal cancer cells in later stages of colorectal carcinogenesis, we developed human colorectal cancer cell lines in which c-Src levels and activity could be inducibly increased by a tightly controlled expression of wild-type c-Src or of the constitutively active mutant of c-Src, c-SrcY527F. Src induction activated multiple signaling pathways (often associated with a proliferative response) but promoted neither cell proliferation in vitro nor tumor growth in a xenograft model in vivo. These results indicate that, in more advanced stages of colorectal carcinogenesis, increases in c-Src levels and activity are likely to have functions other than the direct promotion of tumor growth.
Sigma-Aldrich
Anti-v-Src (Ab-1) Mouse mAb (327), liquid, clone 327, Calbiochem®