Merck
  • Bilirubin: a natural inhibitor of vascular smooth muscle cell proliferation.
Bilirubin: a natural inhibitor of vascular smooth muscle cell proliferation.
Circulation (2005-08-10)
Robert Ollinger, Martin Bilban, Anna Erat, Alberto Froio, James McDaid, Shivraj Tyagi, Eva Csizmadia, Aurelio V Graça-Souza, Angela Liloia, Miguel P Soares, Leo E Otterbein, Anny Usheva, Kenichiro Yamashita, Fritz H Bach
ABSTRACT
Bilirubin, a natural product of heme catabolism by heme oxygenases, was considered a toxic waste product until 1987, when its antioxidant potential was recognized. On the basis of observations that oxidative stress is a potent trigger in vascular proliferative responses, that heme oxygenase-1 is antiatherogenic, and that several studies now show that individuals with high-normal or supranormal levels of plasma bilirubin have a lesser incidence of atherosclerosis-related diseases, we hypothesized that bilirubin would have salutary effects on preventing intimal hyperplasia after balloon injury. We found less balloon injury-induced neointima formation in hyperbilirubinemic Gunn rats and in wild-type rats treated with biliverdin, the precursor of bilirubin, than in controls. In vitro, bilirubin and biliverdin inhibited serum-driven smooth muscle cell cycle progression at the G1 phase via inhibition of the mitogen-activated protein kinase signal transduction pathways and inhibition of phosphorylation of the retinoblastoma tumor suppressor protein. Bilirubin and biliverdin might be potential therapeutics in vascular proliferative disorders.
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)