Merck
  • Ciliary localization of folliculin mediated via a kinesin-2-binding motif is required for its functions in mTOR regulation and tumor suppression.
Ciliary localization of folliculin mediated via a kinesin-2-binding motif is required for its functions in mTOR regulation and tumor suppression.
FEBS letters (2020-10-17)
Yunlong Zhang, Ying Liu, Yu Dai, Yazhe Ren, Guangsen Bao, Bo Ai, Yu Jiang
ABSTRACT
Folliculin (FLCN) is a tumor suppressor protein involved in many cellular processes, including cell signaling, apoptosis, and autophagy. In ciliated cells, FLCN localizes to primary cilia and controls mTORC1 signaling in response to flow stress. Here, we show that the ciliary localization of FLCN requires its interaction with kinesin-2, the motor protein for anterograde intraflagellar transport. FLCN binds to kinesin-2 through a loop region in the middle of the protein. Single point mutations within this region of FLCN disrupt its kinesin-2 binding and ciliary entry. The mutants lose the ability to suppress the abnormal mTORC1/2 signaling activities and anchorage-independent growth of FLCN-deficient tumor cells. These observations suggest that ciliary localization of FLCN is essential for its function as a tumor suppressor.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)