Merck
  • Silsesquioxane Derivatives as Functional Additives for Preparation of Polyethylene-Based Composites: A Case of Trisilanol Melt-Condensation.
Silsesquioxane Derivatives as Functional Additives for Preparation of Polyethylene-Based Composites: A Case of Trisilanol Melt-Condensation.
Polymers (2020-10-08)
Dariusz Brząkalski, Robert E Przekop, Bogna Sztorch, Paulina Jakubowska, Marek Jałbrzykowski, Bogdan Marciniec
ABSTRACT
In this work, polyethylene (PE) composites were prepared with a series of completely condensed silsesquioxanes (SSQ), as well as with open-cage hepta(isobutyl)trisilanol silsesquioxane. The effect of the additives on the thermal, mechanical, rheological, and crystalline properties of the composites obtained was determined. The dispersion of trisilanol derivative within polymer matrix was slightly better than that of the other isobutyl compounds, suggesting condensation of the additive to less polar products of different structure, which was confirmed by thermogravimetry (TG) and matrix-assisted laser desorption-ionization time-of-flight (MALDI-TOF) mass spectrometry analysis. The additives improved the thermal stability of polyethylene and formed composites of higher rigidity than the neat polyolefin. The results were compared to the literature data, with aminopropylhepta(isobutyl)silsesquioxane and vinylhepta(isobutyl)silsesquioxane being used partially as references, as PE composites thereof were reported earlier, but lacked some analytical results and required further investigation. It was proven that the practical upper loading limit for such silsesquioxane compounds as processing and functional additives for polyethylene should be fixed at around 1%.
Sigma-Aldrich
Trichlorovinylsilane, 97%
Sigma-Aldrich
(3-Chloropropyl)triethoxysilane, 95%