Merck
  • Protocols for the Analysis of microRNA Expression, Biogenesis, and Function in Immune Cells.
Protocols for the Analysis of microRNA Expression, Biogenesis, and Function in Immune Cells.
Current protocols in immunology (2019-09-05)
Nannan Zhang, Guowu Hu, Timothy G Myers, Peter R Williamson
ABSTRACT
MicroRNAs (miRNAs) are short (19- to 25-nucleotide) noncoding RNA molecules that target mRNAs to repress gene expression and that play important roles in regulating many fundamental biological functions including cell differentiation, development, growth, and metabolism. They are well conserved in eukaryotic cells and are considered essential ancient elements of gene regulation. miRNA genes are transcribed by RNA polymerase II to generate primary miRNAs (pri-miRNAs), which are cleaved by microprocessor complex in the nucleus to generate stem-loop structures known as pre-miRNAs. Pre-miRNAs are translocated to the cytoplasm and cleaved by Dicer to form the mature miRNAs, which mediate mRNA degradation through their loading to the RNA-induced silencing complex (RISC) and binding to complementary sequences within target mRNAs to repress their translation by mRNA degradation and/or translation inhibition. Because ∼1900 miRNA genes are reported in the human genome, many associated with disease, appropriate methods to study miRNA expression and regulation under physiological and pathological conditions have become increasingly important to the study of many aspects of human biology, including immune regulation. As with small interfering RNA (siRNA), the mechanism of miRNA-mediated targeting has been used to develop miRNA-based therapeutics. For a complete and systematic analysis, it is critical to utilize a variety of different tools to analyze the expression of pri-mRNAs, pre-miRNAs, and mature miRNAs and characterize their targets both in vitro and in vivo. Such studies will facilitate future novel drug design and development. This unit provides six basic protocols for miRNA analysis, covering next-generation sequencing, quantitative real-time PCR (qRT-PCR), and digoxigenin-based expression analysis of pri-mRNAs, pre-miRNAs, and mature miRNAs; mapping of pri-miRNA and their cleavage sites by rapid amplification of cDNA ends (RACE); electrophoretic mobility shift assays (EMSAs) or biotin-based nonradioactive detection of miRNA-protein complexes (miRNPs); and functional analysis of miRNAs using miRNA mimics and inhibitors. © 2019 by John Wiley & Sons, Inc.
Sigma-Aldrich
RIPAb+ pan Ago - RIP Validated Antibody and Primer Set, clone 2A8, from mouse