Merck
  • Interleukin-34 contributes to poor prognosis in triple-negative breast cancer.
Interleukin-34 contributes to poor prognosis in triple-negative breast cancer.
Breast cancer (Tokyo, Japan) (2020-06-25)
Nabeel Kajihara, Fumihito Kitagawa, Takuto Kobayashi, Haruka Wada, Ryo Otsuka, Ken-Ichiro Seino
ABSTRACT
Triple-negative breast cancer (TNBC) is a subtype characterized by the absence of therapeutic targets. It shows rapid progression, higher relapse, and poor prognosis, so the establishment of an effective therapeutic target is required. We focused on interleukin-34 (IL-34) that is a novel cytokine relating to inflammation and tumorigenesis. It has been reported that IL-34 correlates with poor prognosis of various cancers. In this study, we evaluated the relationship of IL-34 and prognosis in TNBC using human clinical information and mice model. We found that IL-34 was highly expressed in TNBC, and the survival rate in TNBC was significantly lower in patients with high IL-34 expression. Furthermore, multivariate analysis revealed that IL-34 independently affects prognosis. In murine TNBC model, IL-34 deficiency in tumor cells decreased in vivo tumor growth and increased inflammatory cytokine production from macrophages. These results suggest that tumor-derived IL-34 creates a favorable environment for TNBC cells. Thus, we showed a novel pathological role of IL-34 in TNBC and the potential of IL-34 as a therapeutic target for it.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Human IL34 / Interleukin-34 ELISA Kit