Merck
Liposomal Nanotherapy for Treatment of Atherosclerosis.
Advanced healthcare materials (2020-06-17)
Anastasia Darwitan, Yee Shan Wong, Luong T H Nguyen, Bertrand Czarny, Anita Vincent, Anu Maashaa Nedumaran, Yang Fei Tan, Aristo Muktabar, Jin Kai Tang, Kee Woei Ng, Subbu Venkatraman
ABSTRACT
Atherosclerosis is a chronic disease that can lead to life-threatening events such as myocardial infarction and stroke, is characterized by the build-up of lipids and immune cells within the arterial wall. It is understood that inflammation is a hallmark of atherosclerosis and can be a target for therapy. In support of this concept, an injectable nanoliposomal formulation encapsulating fluocinolone acetonide (FA), a corticosteroid, is developed that allows for drug delivery to atherosclerotic plaques while reducing the systemic exposure to off-target tissues. In this study, FA is successfully incorporated into liposomal nanocarriers of around 100 nm in size with loading efficiency of 90% and the formulation exhibits sustained release up to 25 d. The anti-inflammatory effect and cholesterol efflux capability of FA-liposomes are demonstrated in vitro. In vivo studies carried out with an apolipoprotein E-knockout (Apoe-/- ) mouse model of atherosclerosis show accumulation of liposomes in atherosclerotic plaques, colocalization with plaque macrophages and anti-atherogenic effect over 3 weeks of treatment. This FA-liposomal-based nanocarrier represents a novel potent nanotherapeutic option for atherosclerosis.
Sigma-Aldrich
Cholesterol Efflux Assay Kit, Sufficient for 100 Fluorometric tests