Merck
  • Examine growth inhibition pattern and lactic acid production in Streptococcus mutans using different concentrations of xylitol produced from Candida tropicalis by fermentation.
Examine growth inhibition pattern and lactic acid production in Streptococcus mutans using different concentrations of xylitol produced from Candida tropicalis by fermentation.
Anaerobe (2012-03-24)
Swati Misra, Shailendra Raghuwanshi, Pritesh Gupta, R K Saxena
ABSTRACT
Twenty clinical isolates of Streptococcus sp. were isolated from six clinical samples of dental caries on MSFA. Amongst these isolates, five clinical isolates were identified as S treptococcus mutans on the basis of morphological, biochemical and 16S rDNA sequencing. The isolated strains of S. mutans were exposed to fermented and purified xylitol (0.25-15.0%) and tested for its anti-microbial effects against control medium (Brain Heart Infusion without xylitol) after 12 h. The plate assay was developed using bromocresol green as an indicator dye in order to study the relative growth inhibition pattern of clinical sample at different concentrations of an anti-microbial compound in a single petriplate. The morphology of S. mutans cells in brain heart infusion (BHI) medium containing xylitol resulted in a diffused cell wall as observed using gram staining technique. The minimum inhibitory concentration (MIC) is 0.25% for S. mutans obtained from different clinical samples. The MIC(50) and MIC(90) is 5.0% and 10.0% xylitol respectively of the selected S. mutans being designated as clinical isolate B (6). The zone of inhibition was 72 mm and lactic acid production was 0.010 g/l at 10% xylitol concentration in Brain Heart Infusion Broth.