Merck
  • Clusterin is a secreted marker for a hypoxia-inducible factor-independent function of the von Hippel-Lindau tumor suppressor protein.
Clusterin is a secreted marker for a hypoxia-inducible factor-independent function of the von Hippel-Lindau tumor suppressor protein.
The American journal of pathology (2006-01-27)
Eijiro Nakamura, Paula Abreu-e-Lima, Yasuo Awakura, Takahiro Inoue, Toshiyuki Kamoto, Osamu Ogawa, Hirokazu Kotani, Toshiaki Manabe, Guo-Jun Zhang, Keiichi Kondo, Vānia Nosé, William G Kaelin
ABSTRACT
Germline mutations in the von Hippel-Lindau (VHL) tumor suppressor gene predispose people to renal cancer, hemangioblastomas, and pheochromocytomas in an allele-specific manner. The best documented function of the VHL gene product (pVHL) relates to its ability to polyubiquitinate, and hence target for destruction, the alpha subunits of the heterodimeric transcription factor hypoxia-inducible factor (HIF). pVHL mutants linked to familial pheochromocyctoma (type 2C VHL disease), in contrast to classical VHL disease, appear to be normal with respect to HIF regulation. Using a simple method for identifying proteins that are differentially secreted by isogenic cell line pairs, we confirmed that the HIF targets IGBP3 and PAI-1 are overproduced by pVHL-defective renal carcinoma cells. In addition, cells lacking wild-type pVHL, including cells producing type 2C pVHL mutants, were defective with respect to expression and secretion of clusterin, which does not behave like a HIF target. Decreased clusterin secretion by pVHL-defective tumors was confirmed in vivo by immunohistochemistry. Therefore, clusterin is a secreted marker for a HIF-independent pVHL function that might be especially important in pheochromocytoma development.
Sigma-Aldrich
Anti-Clusterin α chain (human) Antibody, clone 41D, clone 41D, Upstate®, from mouse
Sign Into View Organizational & Contract Pricing