Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Serine protease inhibitor nafamostat given before reperfusion reduces inflammatory myocardial injury by complement and neutrophil inhibition.
Serine protease inhibitor nafamostat given before reperfusion reduces inflammatory myocardial injury by complement and neutrophil inhibition.
Journal of cardiovascular pharmacology (2008-08-02)
Hansjörg Schwertz, Justin M Carter, Martin Russ, Sebastian Schubert, Axel Schlitt, Ute Buerke, Martin Schmidt, Heinz Hillen, Karl Werdan, Michael Buerke
ABSTRACT
Animal data strongly support a role for inflammation in myocardial ischemia reperfusion injury. Attempts at cardioprotection by immunomodulation (such as with the specific C5 antibody pexelizumab) in humans have been disappointing. We hypothesized that a broader spectrum antiinflammatory agent might yield successful cardioprotection. The serine protease inhibitor nafamostat (FUT-175), which is already in clinical use, is a potent antiinflammatory synthetic serine protease inhibitor with anticomplement activity that we tested in a well-established rabbit model of 1 hour of myocardial ischemia followed by 3 hours of reperfusion. Compared to vehicle-treated animals, nafamostat (1 mg/kg of body weight) administered 5 minutes before reperfusion significantly reduced myocardial injury assessed by plasma creatine kinase activity (38.1 +/- 6.0 versus 57.9 +/- 3.7I U/g protein; P < 0.05) and myocardial necrosis (23.6 +/- 3.1% versus 35.7 +/- 1.0%; P < 0.05) as well as myocardial leukocyte accumulation (P < 0.05). In parallel in vitro studies, Nafamostat was a significantly more potent broad spectrum complement suppressor than C1 inhibitor. Nafamostat appears to have capability as an inhibitor of both complement pathways and as a broad-spectrum antiinflammatory agent by virtue of its serine protease inhibition. Administration of nafamostat before myocardial reperfusion after ischemia produced significant, dose-dependent cardioprotection. Reduced leukocyte accumulation and complement activity seem involved in the mechanism of this cardioprotective effect.
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein