Merck
  • Systemic administration of GMP induces anxiolytic-like behavior in rats.
Systemic administration of GMP induces anxiolytic-like behavior in rats.
Pharmacology, biochemistry, and behavior (2010-06-23)
Roberto Farina Almeida, Victor Hermes Cereser, Rafael Berger Faraco, Ana Elisa Böhmer, Diogo Onofre Souza, Marcelo Ganzella
ABSTRACT
The glutamatergic system has received considerable attention over the last few years as potential target to develop anxiolytic drugs. Guanine based purines (GBPs) play an important neurmodulatory effect in the glutamatergic system. Several studies have shown the ability of the GBPs to reduce glutamatergic activity. In the present study, we investigated the anxiolytic effect of GBPs - by Guanosina Monophosphate (GMP) administration - in rodents. Adult male Wistar rats were pretreated with GMP (10, 25, 50, 100 and 150mg/kg: i.p.); or saline (NaCl 0.9%; i.p.) (control); or, diazepam (2mg/kg: i.p.) (positive control). One hour after the injection, the anxiety-related behaviors for each animal was evaluated in the light/dark, elevated plus-maze, and open field tasks. Additionally, purines concentration in the cerebrospinal fluid (CSF) was verified. The administration of 25 and 50mg/kg GMP was able to promote anxiolytic-like behavior, in the light/dark and elevated plus-maze task, similar to diazepam effect. However, no changes in the open field task, or CSF purines concentration were found for either GMP or diazepam treated animals, when compared with saline group. Thus, this study suggests that acute administration of GMP was able to decrease the levels of anxiety in classical behavioral tasks.
Sigma-Aldrich
Guanosine 5′-monophosphate disodium salt hydrate, from yeast, ≥99%