Merck
  • Cytotoxic phenanthroline derivatives alter metallostasis and redox homeostasis in neuroblastoma cells.
Cytotoxic phenanthroline derivatives alter metallostasis and redox homeostasis in neuroblastoma cells.
Oncotarget (2018-12-18)
Irina Naletova, Cristina Satriano, Alessandra Curci, Nicola Margiotta, Giovanni Natile, Giuseppe Arena, Diego La Mendola, Vincenzo Giuseppe Nicoletti, Enrico Rizzarelli
ABSTRACT
Copper homeostasis is generally investigated focusing on a single component of the metallostasis network. Here we address several of the factors controlling the metallostasis for neuroblastoma cells (SH-SY5Y) upon treatment with 2,9-dimethyl-1,10-phenanthroline-5,6-dione (phendione) and 2,9-dimethyl-1,10-phenanthroline (cuproindione). These compounds bind and transport copper inside cells, exert their cytotoxic activity through the induction of oxidative stress, causing apoptosis and alteration of the cellular redox and copper homeostasis network. The intracellular pathway ensured by copper transporters (Ctr1, ATP7A), chaperones (CCS, ATOX, COX 17, Sco1, Sco2), small molecules (GSH) and transcription factors (p53) is scrutinised.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, HRP conjugate, Species Adsorbed, 0.8 mg/mL, Chemicon®
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®