Merck
  • Phospholipase D2 in prostate cancer: protein expression changes with Gleason score.
Phospholipase D2 in prostate cancer: protein expression changes with Gleason score.
British journal of cancer (2019-11-02)
Amanda R Noble, Karen Hogg, Rakesh Suman, Daniel M Berney, Sylvain Bourgoin, Norman J Maitland, Martin G Rumsby
ABSTRACT
Phospholipases D1 and D2 (PLD1/2) are implicated in tumorigenesis through their generation of the signalling lipid phosphatidic acid and its downstream effects. Inhibition of PLD1 blocks prostate cell growth and colony formation. Here a role for PLD2 in prostate cancer (PCa), the major cancer of men in the western world, is examined. PLD2 expression was analysed by immunohistochemistry and western blotting. The effects of PLD2 inhibition on PCa cell viability and cell motility were measured using MTS, colony forming and wound-healing assays. PLD2 protein is expressed about equally in luminal and basal prostate epithelial cells. In cells from different Gleason-scored PCa tissue PLD2 protein expression is generally higher than in non-tumorigenic cells and increases in PCa tissue scored Gleason 6-8. PLD2 protein is detected in the cytosol and nucleus and had a punctate appearance. In BPH tissue stromal cells as well as basal and luminal cells express PLD2. PLD2 protein co-expresses with chromogranin A in castrate-resistant PCa tissue. PLD2 inhibition reduces PCa cell viability, colony forming ability and directional cell movement. PLD2 expression correlates with increasing Gleason score to GS8. PLD2 inhibition has the potential to reduce PCa progression.
Sigma-Aldrich
Pyrimidine, ≥98.0%
Sigma-Aldrich
VU0155056, ≥98% (HPLC)