Merck
  • MicroRNA-377 exerts a potent suppressive role in osteosarcoma through the involvement of the histone acetyltransferase 1-mediated Wnt axis.
MicroRNA-377 exerts a potent suppressive role in osteosarcoma through the involvement of the histone acetyltransferase 1-mediated Wnt axis.
Journal of cellular physiology (2019-06-04)
Peng Xia, Rui Gu, Wei Zhang, Liwei Shao, Fang Li, Changyan Wu, Yifu Sun
ABSTRACT
It has been demonstrated that microRNAs (miRNAs) may contribute to tumorigenesis and tumor growth in osteosarcoma (OS), which is a primary malignant tumor of bone frequently diagnosed in adolescents and young people. The purpose of our investigation was to evaluate the functional relevance of miR-377 in OS and to investigate whether the mechanism was related to the histone acetyltransferase 1 (HAT1)-mediated Wnt signaling pathway. By screening differentially expressed genes in microarray GSE47572, HAT1 was found to be a candidate gene of interest. Besides, the regulatory miRNA (miR-377) of HAT1 was also selected. The interaction among miR-377, HAT1, and the Wnt signaling pathway was evaluated. In addition, the miR-377 expression was altered in OS cells (U-2OS and SOSP-9607) to assess the in vitro cell apoptosis and the in vivo tumor growth. OS tissues presented elevated HAT1 expression and decreased miR-377 expression. A putative miR-377 binding site in HAT1 3'-UTR HAT1 was verified. Cells with miR-377 overexpression or HAT1 silencing were observed to exhibit reduced HAT1 expression and promoted apoptosis, accompanied by blockade of Wnt signaling. Moreover, the in vivo experiment revealed that miR-377 overexpression or HAT1 silencing inhibited tumor growth and reduced tumor size in nude mice. Taken together, our results conclude that miR-377 may promote OS cell apoptosis through inactivation of the HAT1-mediated Wnt signaling pathway, highlighting the potential therapeutic effect of miR-377 on OS treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human HAT1