Merck
  • Downregulation of microRNA-129-5p increases the risk of intervertebral disc degeneration by promoting the apoptosis of nucleus pulposus cells via targeting BMP2.
Downregulation of microRNA-129-5p increases the risk of intervertebral disc degeneration by promoting the apoptosis of nucleus pulposus cells via targeting BMP2.
Journal of cellular biochemistry (2019-08-23)
Weijie Yang, Ping Sun
ABSTRACT
miR-129-5p is implicated in many diseases, such as laryngeal cancer and breast cancer. In this study, we studied the mechanism underlying the role of BMP2 in intervertebral disc degeneration (IDD). We used a luciferase assay system to determine the relationship between BMP2 and miR-129-5 expression. In addition, Western blot and real-time PCR were used to confirm the regulatory relationship between miR-129-5p and its targets, while flow cytometry was used to evaluate the effect of miR-129-5p on the apoptosis of neural progenitor cells (NPCs). BMP2 was confirmed as a direct target of miR-129-5p. Furthermore, the expression of miR-129 was downregulated along with upregulated BMP2 expression in IDD patients. Meanwhile, BMP2 was validated as the target of miR-129-5p in cells transfected with miR-129-5p and BMP2 siRNA. Also, compared with NPCs transfected with blank/scramble controls or miR-129-5p inhibitors, the NPCs treated with miR-129-5p mimics or BMP2 siRNA exhibited evidently elevated viability and inhibited apoptosis. The data demonstrated that miR-129-5p was poorly expressed in IDD patients, and the dysregulation of miR-129-5p might contribute to the development of IDD by targeting BMP2 expression.
Sigma-Aldrich
MISSION® esiRNA, targeting human BMP2