Merck
  • Antagonism of the effects of (+)-PD 128907 on midbrain dopamine neurones in rat brain slices by a selective D2 receptor antagonist L-741,626.
Antagonism of the effects of (+)-PD 128907 on midbrain dopamine neurones in rat brain slices by a selective D2 receptor antagonist L-741,626.
British journal of pharmacology (1996-12-01)
B J Bowery, Z Razzaque, F Emms, S Patel, S Freedman, L Bristow, J Kulagowski, G R Seabrook
ABSTRACT
1. The ability of PD 128907 to activate dopamine receptors in the ventral tegmental area, substantia nigra pars compacta, and striatum was investigated by use of in vitro electrophysiological recording and fast cyclic voltammetry. The affinity of a novel D2 selective antagonist L-741,626 for receptors activated by this agonist was measured to determine if its effects were mediated by D2 or D3 receptors. 2. The active (+) enantiomer of PD 128907 bound with high affinity and selectivity to rat D3 dopamine receptors. The Ki values for (+)-PD 128907 were 620 nM at D2, 1 nM at D3 and 720 nM at D4 receptors. 3. (+)-PD 128907 inhibited cell firing in both the ventral tegmental area and substantia nigra pars compacta with EC50 values of 33 nM (pEC50 = 7.48 +/- 0.10, n = 10) and 38 nM (pEC50 = 7.42 +/- 0.15, n = 5), respectively. No effects of (+)-PD 128907 (100 nM) were observed on glutamate or GABA-mediated synaptic potentials elicited by focal bipolar stimulation. 4. L-741,626 antagonized these effects of (+)-PD 128907 in a concentration-dependent and surmountable manner with an affinity, determined from Schild analysis, of 20 nM (pKB = 7.71 +/- 0.14) in the ventral tegmental area and 11 nM (pKB = 7.95 +/- 0.18) in the substantia nigra pars compacta. 5. (+)-PD 128907 also inhibited dopamine release in the caudate-putamen with an EC50 of 66 nM (n = 5). The affinity of L-741,626 for these nerve terminal autoreceptors (pKB = 7.71 +/- 0.06; = 20 nM) was identical to that observed on midbrain dopamine neurones. 6. These data demonstrate that the D3 receptor ligand (+)-PD 128907 is a potent agonist on rat midbrain dopamine neurones. However, its lack of regional selectivity, and the high affinity of the selective D2 receptor antagonist L-741,626 for receptors activated by (+)-PD 128907, was more consistent with an action on D2 autoreceptors rather than upon a D3 dopamine receptor subtype.
Sigma-Aldrich
L-741,626, ≥98% (HPLC)