Merck
  • Essential Gene Profiles for Human Pluripotent Stem Cells Identify Uncharacterized Genes and Substrate Dependencies.
Essential Gene Profiles for Human Pluripotent Stem Cells Identify Uncharacterized Genes and Substrate Dependencies.
Cell reports (2019-04-11)
Barbara Mair, Jelena Tomic, Sanna N Masud, Peter Tonge, Alexander Weiss, Matej Usaj, Amy Hin Yan Tong, Jamie J Kwan, Kevin R Brown, Emily Titus, Michael Atkins, Katherine S K Chan, Lise Munsie, Andrea Habsid, Hong Han, Marion Kennedy, Brenda Cohen, Gordon Keller, Jason Moffat
ABSTRACT
Human pluripotent stem cells (hPSCs) provide an invaluable tool for modeling diseases and hold promise for regenerative medicine. For understanding pluripotency and lineage differentiation mechanisms, a critical first step involves systematically cataloging essential genes (EGs) that are indispensable for hPSC fitness, defined as cell reproduction in this study. To map essential genetic determinants of hPSC fitness, we performed genome-scale loss-of-function screens in an inducible Cas9 H1 hPSC line cultured on feeder cells and laminin to identify EGs. Among these, we found FOXH1 and VENTX, genes that encode transcription factors previously implicated in stem cell biology, as well as an uncharacterized gene, C22orf43/DRICH1. hPSC EGs are substantially different from other human model cell lines, and EGs in hPSCs are highly context dependent with respect to different growth substrates. Our CRISPR screens establish parameters for genome-wide screens in hPSCs, which will facilitate the characterization of unappreciated genetic regulators of hPSC biology.
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.