Merck
  • Transcription Factor NFAT5 Promotes Migration and Invasion of Rheumatoid Synoviocytes via Coagulation Factor III and CCL2.
Transcription Factor NFAT5 Promotes Migration and Invasion of Rheumatoid Synoviocytes via Coagulation Factor III and CCL2.
Journal of immunology (Baltimore, Md. : 1950) (2018-05-26)
Saseong Lee, Jin-Sun Kong, Sungyong You, H Moo Kwon, Seung-Ah Yoo, Chul-Soo Cho, Wan-Uk Kim
ABSTRACT
Fibroblast-like synoviocytes (FLSs) play a key role in the progression of rheumatoid arthritis (RA) as a primary component of invasive hypertrophied pannus. FLSs of RA patients (RA-FLSs) exhibit cancer-like features, including promigratory and proinvasive activities that largely contribute to joint cartilage and bone destruction. In this study, we hypothesized that the NF of activated T cell 5 (NFAT5), a transcription factor involving tumor invasiveness, would control the migration and invasion of RA-FLSs. Analyses of transcriptomes demonstrated the significant involvement of NFAT5 in locomotion of RA-FLSs and that tissue factor (TF; also known as coagulation factor III) and CCL2 were the major downstream target genes of NFAT5 involving FLS migration and invasion. In cultured RA-FLSs, IL-1β and TGF-β increased TF and CCL2 expression by upregulating NFAT5 expression via p38 MAPK. Functional assays demonstrated that NFAT5- or TF-deficient RA-FLSs displayed decreased lamellipodia formation, cell migration, and invasion under IL-1β- or TGF-β-stimulated conditions. Conversely, factor VIIa, a specific activator of TF, increased migration of RA-FLSs, which was blocked by NFAT5 knockdown. Recombinant CCL2 partially restored the decrease in migration and invasion of NFAT5-deficient RA-FLSs stimulated with IL-1β. NFAT5-knockout mouse FLSs also showed decreased expressions of TF and CCL2 and reduced cell migration. Moreover, KRN2, a specific inhibitor of NFAT5, suppressed migration of FLSs stimulated with TGF-β. Conclusively, to our knowledge, this is the first study to provide evidence of a functional link between osmoprotective NFAT5 and TF in the migration and invasion of RA-FLSs and supports a role for NFAT5 blockade in the treatment of RA.